4-Hydroxyphenylboronsäure zur Synthese, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
M-Clarity™ Qualitätsstufe = MQ200
8.43854.0001
8.43854.0005
8.43854.9999
8.43854.0001EA
61.4
EUR
8.43854.0001
8.43854.0005
8.43854.9999
4-Hydroxyphenylboronsäure zur Synthese, Sigma-Aldrich®
4-Hydroxyphenylboronsäure
Formel:
C₆H₇BO₃ Molecular Weight: 137.93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074628 CAS-Nummer: 71597-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...