4-Bromphenylboronsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(4-bromphenyl)boran, 4-Brombenzolboronsäure, (4-Bromphenyl)metaborsäure
L01565.03
L01565.06
L01565.14
L01565.03EA
26.9
EUR
L01565.03
L01565.06
L01565.14
4-Bromphenylboronsäure ≥97%
4-Bromphenylboronsäure
Purity: 97+%
CAS No.: 5467-74-3
Molecular Formula: C6H6BBrO2
Formula Weight: 200.83
Melting Point: 282-286°
Beilstein Reference: 2936347
Formel:
C₆H₆BBrO₂ Molecular Weight: 200,83 g/mol Schmelzpunkt: 282…286 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002104 CAS-Nummer: 5467-74-3 EINECS: 226-779-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...