4-Bromphenylboronsäure
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
Dihydroxy(4-bromphenyl)boran, 4-Brombenzolboronsäure, (4-Bromphenyl)metaborsäure
FL10981.POR
010981-25G
010981-5G
CHMPFL10981.POREA
0
EUR
CHMPFL10981.POR
FLUO010981-25G
FLUO010981-5G
4-Bromphenylboronsäure
4-Bromphenylboronsäure
Formel:
C₆H₆BBrO₂ Molecular Weight: 200,83 g/mol Dichte: 1,67 g/cm³ (20 °C) |
MDL:
MFCD00002104 CAS-Nummer: 5467-74-3 EINECS: 226-779-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...