2-Brom-1,3,5-trichlorbenzol

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Bromo-2,4,6-trichlorobenzene

TL00687.1 TL00687.10 TL00687.50
MAYBTL00687.1EA 0 EUR
MAYBTL00687.1 MAYBTL00687.10 MAYBTL00687.50
2-Brom-1,3,5-trichlorbenzol
2-Brom-1,3,5-trichlorbenzol
Formel: C₆H₂BrCl₃
Molecular Weight: 260,34 g/mol
MDL: MFCD00060653
CAS-Nummer: 19393-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR