2-Brom-4-chlorbenzoesäure

Lieferant: CHEMPUR

B2573-B2.5 018254-1G
CHMPB2573-B2.5EA 0 EUR
CHMPB2573-B2.5 FLUO018254-1G
2-Brom-4-chlorbenzoesäure
2-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00672930
CAS-Nummer: 936-08-3
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR