4-Brom-2-chlorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

664014-25G 664014-5G
SIAL664014-25GEA 67.8 EUR
SIAL664014-25G SIAL664014-5G
4-Brom-2-chlorbenzoesäure, Sigma-Aldrich®
4-Bromo-2-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00040903
CAS-Nummer: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR