4-Brom-3-chlorbenzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

H27252.14 H27252.06
H27252.14EA 232 EUR
H27252.14 H27252.06
4-Brom-3-chlorbenzoesäure ≥97%
4-Bromo-3-chlorobenzoic acid

Purity: 97% CAS No.: 25118-59-6 Molecular Formula: C7H4BrClO2 Formula Weight: 235.47 Odour: Odourless Melting Point: 220-225° MDL No.: MFCD08276864

Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
Schmelzpunkt: 220…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276864
CAS-Nummer: 25118-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR