4-Brom-3-chlorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

665312-25G 665312-5G
SIAL665312-25GEA 173 EUR
SIAL665312-25G SIAL665312-5G
4-Brom-3-chlorbenzoesäure, Sigma-Aldrich®
4-Bromo-3-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD08276864
CAS-Nummer: 25118-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR