1-Brom-2-chlor-5-fluorbenzol

Lieferant: CHEMPUR
Achtung

Synonyme: 2-Bromo-1-chloro-4-fluorobenzene

FL13191.POR 013191-25G 013191-100G 013191-10G
CHMPFL13191.POREA 0 EUR
CHMPFL13191.POR FLUO013191-25G FLUO013191-100G FLUO013191-10G
1-Brom-2-chlor-5-fluorbenzol
1-Brom-2-chlor-5-fluorbenzol
Formel: C₆H₃BrClF
Molecular Weight: 209,44 g/mol
MDL: MFCD00061120
CAS-Nummer: 201849-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR