1-Brom-3-chlor-5-fluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
3-Chloro-5-fluorobromobenzene
004150-25G
004150-100G
FLUO004150-25GEA
34.1
EUR
FLUO004150-25G
FLUO004150-100G
1-Brom-3-chlor-5-fluorbenzol
1-Bromo-3-chloro-5-fluorobenzene
Formel:
C₆H₃BrClF Molecular Weight: 209,44 g/mol |
MDL:
MFCD00070747 CAS-Nummer: 33863-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...