4-Brom-2-chlor-6-fluoranisol

Lieferant: FLUOROCHEM
Achtung

005576-1G 005576-5G
FLUO005576-1GEA 107 EUR
FLUO005576-1G FLUO005576-5G
4-Brom-2-chlor-6-fluoranisol
4-Brom-2-chlor-6-fluoranisol
Formel: C₇H₅BrClFO
Molecular Weight: 239,47 g/mol
MDL: MFCD01631477
CAS-Nummer: 261762-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR