4-Brom-3-chloranisol

Lieferant: FLUOROCHEM

021885-5G 021885-25G
FLUO021885-5GEA 49.1 EUR
FLUO021885-5G FLUO021885-25G
4-Brom-3-chloranisol
4-Brom-3-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
MDL: MFCD00070742
CAS-Nummer: 50638-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR