3-Brom-4-chloranisol ≥98%

Lieferant: Thermo Scientific

Synonyme: 2-Bromo-1-chloro-4-methoxybenzene

A18660.03 A18660.06
A18660.03EA 21.7 EUR
A18660.03 A18660.06
3-Brom-4-chloranisol ≥98%
3-Brom-4-chloranisol

Purity: 98% CAS No.: 2732-80-1 Molecular Formula: C7H6BrClO Formula Weight: 221.48 MDL No.: MFCD00070739

Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070739
CAS-Nummer: 2732-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR