4-Brombenzhydrazid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Brombenzoesäurehydrazid, 4-Brombenzohydrazid, 4-Brombenzol-1-carbohydrazid
A18918.06
A18918.14
A18918.06EA
33.3
EUR
A18918.06
A18918.14
4-Brombenzhydrazid ≥98%
4-Brombenzhydrazid
Purity: 98+%
CAS No.: 5933-32-4
Molecular Formula: C7H7BrN2O
Formula Weight: 215.05
Melting Point: 167-169°
Beilstein Reference: 777006
Formel:
C₇H₇BrN₂O Molecular Weight: 215,05 g/mol Schmelzpunkt: 167…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007602 CAS-Nummer: 5933-32-4 EINECS: 227-681-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...