4-Brombenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
4-Brombenzoesäurehydrazid, 4-Brombenzohydrazid, 4-Brombenzol-1-carbohydrazid
018244-5G
018244-25G
FLUO018244-5GEA
48.8
EUR
FLUO018244-5G
FLUO018244-25G
4-Brombenzhydrazid
4-Brombenzhydrazid
Formel:
C₇H₇BrN₂O Molecular Weight: 215,05 g/mol |
MDL:
MFCD00007602 CAS-Nummer: 5933-32-4 EINECS: 227-681-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...