Ethyl-2-brombenzoat
Lieferant: Thermo Scientific
Synonyme:
2-Brombenzoesäureethylester
RDP00160.1
RDP00160EA
RDP00160EE
MAYBRDP00160.1EA
0
EUR
MAYBRDP00160.1
MAYBRDP00160EA
MAYBRDP00160EE
Ethyl-2-brombenzoat
Ethyl-2-brombenzoat
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol Siedepunkt: 265 °C (1013 hPa) Dichte: 1,441 g/cm³ (20 °C) |
MDL:
MFCD00015443 CAS-Nummer: 6091-64-1 EINECS: 228-034-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...