Methyl-2-brombenzoat

Lieferant: Thermo Scientific

Synonyme: 2-Brombenzoesäuremethylester

RDP00159.1 RDP00159.50
MAYBRDP00159.1EA 0 EUR
MAYBRDP00159.1 MAYBRDP00159.50
Methyl-2-brombenzoat
Methyl-2-brombenzoat
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
Siedepunkt: 253 °C (1013 hPa)
Dichte: 1,51 g/cm³ (20 °C)
MDL: MFCD00016328
CAS-Nummer: 610-94-6
EINECS: 210-241-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR