4-Amino-5-brom-2-methoxybenzoesäure

Lieferant: BIONET RESEARCH

2R-0683.500 2R-0683.1
BIOT2R-0683.500EA 0 EUR
BIOT2R-0683.500 BIOT2R-0683.1
4-Amino-5-brom-2-methoxybenzoesäure
4-Amino-5-brom-2-methoxybenzoesäure
Formel: C₈H₈BrNO₃
Molecular Weight: 246.06 g/mol
MDL: MFCD02186027
CAS-Nummer: 35290-97-2
EINECS: 252-491-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR