3-Brom-4-hydroxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

699926-25G 699926-5G
SIAL699926-25GEA 212 EUR
SIAL699926-25G SIAL699926-5G
3-Brom-4-hydroxybenzoesäure, Sigma-Aldrich®
3-Bromo-4-hydroxybenzoic acid
Formel: C₇H₅BrO₃
Molecular Weight: 217,02 g/mol
MDL: MFCD00017547
CAS-Nummer: 14348-41-5
EINECS: 238-299-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR