1-Brom-2,3,5-trifluorbenzol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
2,3,5-Trifluorobromobenzene
007834-25G
007834-100G
FLUO007834-25GEA
354
EUR
FLUO007834-25G
FLUO007834-100G
1-Brom-2,3,5-trifluorbenzol
1-Bromo-2,3,5-trifluorobenzene
Formel:
C₆H₂BrF₃ Molecular Weight: 210,98 g/mol |
MDL:
MFCD00012232 CAS-Nummer: 133739-70-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...