Chlorophenol Red-β-D-galactopyranoside ≥90% (durch HPLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
chlorophenol red galactopyranoside, CPRG, 2-Chlor-4-[3-(3-chlor-4-hydroxyphenyl)-1,1-dioxido-3H-2,1-benzoxathiol-3-yl]phenyl-β-D-galactopyranosid
59767-100MG-F
59767-25MG-F
SIAL59767-100MG-FEA
151
EUR
SIAL59767-100MG-F
SIAL59767-25MG-F
Chlorophenol Red-β-D-galactopyranoside ≥90% (durch HPLC), Sigma-Aldrich®
Chlorophenol red-β-D-galactopyranoside (CPRG)
Suitable as substrate for β-galactosidase.
Measures the expression levels of β-galactosidase in transfected cells. The lacZ gene is a popular reporter gene in transfection reactions because its product, β-galactosidase, is a very stable enzyme.
Formel:
C₂₅H₂₂Cl₂O₁₀S Molecular Weight: 585,407 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD05863726 CAS-Nummer: 99792-79-7 |
Spezifikation Testergebnisse
Assay | ≥90% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...