3-Brom-2-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF841.1 005473-1G 005473-5G
CHMPCFF841.1EA 522 EUR
CHMPCFF841.1 FLUO005473-1G FLUO005473-5G
3-Brom-2-fluorbenzoesäure
3-Bromo-2-fluorobenzoic acid
Formel: C₇H₄BrFO₂
MDL: MFCD00665763
CAS-Nummer: 161957-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR