5-Brom-2-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF846.1 005477-1G 005477-5G 005477-25G
CHMPCFF846.1EA 522 EUR
CHMPCFF846.1 FLUO005477-1G FLUO005477-5G FLUO005477-25G
5-Brom-2-fluorbenzoesäure
5-Brom-2-fluorbenzoesäure
Formel: C₇H₄BrFO₂
MDL: MFCD00143423
CAS-Nummer: 146328-85-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR