5-Brom-2-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

636452-1G 636452-5G
SIAL636452-1GEA 30 EUR
SIAL636452-1G SIAL636452-5G
5-Brom-2-fluorbenzoesäure, Sigma-Aldrich®
5-Brom-2-fluorbenzoesäure
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00143423
CAS-Nummer: 146328-85-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR