4-Brom-2,6-difluorbenzoesäure

Lieferant: CHEMPUR
Achtung

FL13022.POR 013022-5G 013022-1G 013022-25G
CHMPFL13022.POREA 0 EUR
CHMPFL13022.POR FLUO013022-5G FLUO013022-1G FLUO013022-25G
4-Brom-2,6-difluorbenzoesäure
4-Bromo-2,6-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
MDL: MFCD03094085
CAS-Nummer: 183065-68-1
EINECS: 457-940-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR