4-Brom-3-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF845.1 005476-1G 005476-5G 005476-25G
CHMPCFF845.1EA 522 EUR
CHMPCFF845.1 FLUO005476-1G FLUO005476-5G FLUO005476-25G
4-Brom-3-fluorbenzoesäure
4-Bromo-3-fluorobenzoic acid
Formel: C₇H₄BrFO₂
MDL: MFCD00672932
CAS-Nummer: 153556-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR