1-Brom-2-fluor-5-iodbenzol

Lieferant: CHEMPUR
Achtung

FL13233.POR 013233-5G 013233-25G
CHMPFL13233.POREA 0 EUR
CHMPFL13233.POR FLUO013233-5G FLUO013233-25G
1-Brom-2-fluor-5-iodbenzol
1-Brom-2-fluor-5-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
MDL: MFCD04116322
CAS-Nummer: 811842-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR