4-Brom-3-fluorphenylisothiocyanat

Lieferant: FLUOROCHEM

009605-1G 009605-5G
FLUO009605-1GEA 168 EUR
FLUO009605-1G FLUO009605-5G
4-Brom-3-fluorphenylisothiocyanat
4-Brom-3-fluorphenylisothiocyanat
Formel: C₇H₃BrFNS
Molecular Weight: 232,08 g/mol
CAS-Nummer: 362690-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR