3-Brom-4-fluorphenylisothiocyanat

Lieferant: FLUOROCHEM

023857-1G 023857-5G
FLUO023857-1GEA 303 EUR
FLUO023857-1G FLUO023857-5G
3-Brom-4-fluorphenylisothiocyanat
3-Brom-4-fluorphenylisothiocyanat
Formel: C₇H₃BrFNS
Molecular Weight: 232,08 g/mol
CAS-Nummer: 710351-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR