2-Brom-4-fluorphenylisothiocyanat

Lieferant: CHEMPUR

FL8215.POR FL19472.5 FL19472.25 008215-25G 008215-5G
CHMPFL8215.POREA 0 EUR
CHMPFL8215.POR CHMPFL19472.5 CHMPFL19472.25 FLUO008215-25G FLUO008215-5G
2-Brom-4-fluorphenylisothiocyanat
2-Brom-4-fluorphenylisothiocyanat
Formel: C₇H₃BrFNS
Molecular Weight: 232,08 g/mol
MDL: MFCD00673194
CAS-Nummer: 175205-35-3
UN: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR