2-Brom-5-fluorphenylisothiocyanat
Lieferant: FLUOROCHEM
009607-1G
009607-5G
FLUO009607-1GEA
45.9
EUR
FLUO009607-1G
FLUO009607-5G
2-Brom-5-fluorphenylisothiocyanat
2-Brom-5-fluorphenylisothiocyanat
Formel:
C₇H₃BrFNS Molecular Weight: 232,08 g/mol |
MDL:
MFCD00276607 CAS-Nummer: 1027513-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...