2,6-Dibrom-4-fluoranilin

Lieferant: Thermo Scientific

BTB04549.1 BTB04549.10 BTB04549.50
MAYBBTB04549.1EA 0 EUR
MAYBBTB04549.1 MAYBBTB04549.10 MAYBBTB04549.50
2,6-Dibrom-4-fluoranilin
2,6-Dibrom-4-fluoranilin
Formel: C₆H₄Br₂FN
Molecular Weight: 268.91 g/mol
Siedepunkt: 277
Schmelzpunkt: 68
MDL: MFCD00041445
CAS-Nummer: 344-18-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR