3-Brom-4-fluoranilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25610.06
B25610.14
B25610.06EA
60.8
EUR
B25610.06
B25610.14
3-Brom-4-fluoranilin ≥98%
3-Brom-4-fluoranilin
Purity: 98%
CAS No.: 656-64-4
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Melting Point: 34-36°
Boiling Point: 235°
UN No.: 2811
MDL No.: MFCD03788559
Formel:
C₆H₅BrFN Molecular Weight: 190.01 g/mol Siedepunkt: 235 °C Schmelzpunkt: 34…36 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788559 CAS-Nummer: 656-64-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...