4-Brom-2,5-difluoranilin

Lieferant: FLUOROCHEM

005236-5G 005236-25G
FLUO005236-5GEA 36.9 EUR
FLUO005236-5G FLUO005236-25G
4-Brom-2,5-difluoranilin
4-Brom-2,5-difluoranilin
Formel: C₆H₄BrF₂N
MDL: MFCD01861125
CAS-Nummer: 112279-60-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR