4-Brom-2,5-difluoranilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

638412-1G 638412-5G
SIAL638412-1GEA 26 EUR
SIAL638412-1G SIAL638412-5G
4-Brom-2,5-difluoranilin, Sigma-Aldrich®
4-Brom-2,5-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208,01 g/mol
MDL: MFCD01861125
CAS-Nummer: 112279-60-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR