4-Brom-2,6-difluoranilin

Lieferant: CHEMPUR

CD04900.250 CD04900.1
CHMPCD04900.250EA 0 EUR
CHMPCD04900.250 MAYBCD04900.1
4-Brom-2,6-difluoranilin
4-Brom-2,6-difluoranilin
Formel: C₆H₄BrF₂N
Molecular Weight: 208.01 g/mol
MDL: MFCD00013389
CAS-Nummer: 67567-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR