4-Brom-2,5-difluoranisol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-Bromo-2,5-difluoro-4-methoxybenzene

004185-5G 004185-25G
FLUO004185-5GEA 31.1 EUR
FLUO004185-5G FLUO004185-25G
4-Brom-2,5-difluoranisol
4-Brom-2,5-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.02 g/mol
MDL: MFCD00143258
CAS-Nummer: 202865-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR