1-Brom-2-(difluormethoxy)benzol

Lieferant: FLUOROCHEM

004835-5G 004835-25G
FLUO004835-5GEA 0 EUR
FLUO004835-5G FLUO004835-25G
1-Brom-2-(difluormethoxy)benzol
1-Brom-2-(difluormethoxy)benzol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.02 g/mol
MDL: MFCD00221468
CAS-Nummer: 175278-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR