5-Brom-2,3-difluoranisol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 5-Bromo-1,2-difluoro-3-methoxybenzene

005866-5G 005866-25G
FLUO005866-5GEA 39.9 EUR
FLUO005866-5G FLUO005866-25G
5-Brom-2,3-difluoranisol
5-Brom-2,3-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.02 g/mol
MDL: MFCD01631352
CAS-Nummer: 261762-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR