2-Brom-4,6-difluoranisol

Lieferant: CHEMPUR
Achtung

CFF830. FL13195.POR 013195-5G
CHMPCFF830.EA 0 EUR
CHMPCFF830. CHMPFL13195.POR FLUO013195-5G
2-Brom-4,6-difluoranisol
2-Brom-4,6-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00070754
CAS-Nummer: 202865-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR