4-Brom-2,6-difluoranisol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 5-Bromo-1,3-difluoro-2-methoxybenzene

005578-5G 005578-10G 005578-1G
FLUO005578-5GEA 177 EUR
FLUO005578-5G FLUO005578-10G FLUO005578-1G
4-Brom-2,6-difluoranisol
4-Brom-2,6-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.02 g/mol
MDL: MFCD01631351
CAS-Nummer: 104197-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR