1-Brom-4-(difluormethoxy)benzol

Lieferant: Thermo Scientific

Synonyme: 4-(Difluoromethoxy)bromobenzene

TL00753.1 TL00753.10
MAYBTL00753.1EA 0 EUR
MAYBTL00753.1 MAYBTL00753.10
1-Brom-4-(difluormethoxy)benzol
1-Brom-4-(difluormethoxy)benzol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00221465
CAS-Nummer: 5905-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR