2-Brom-5-fluoranisol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-fluoro-2-methoxybenzene
H26325.06
H26325.14
H26325.06EA
109
EUR
H26325.06
H26325.14
2-Brom-5-fluoranisol ≥97%
2-Brom-5-fluoranisol
Purity: 97%
CAS No.: 450-88-4
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Flash Point: 90°(194°F)
Refractive Index: 1.5460
Boiling Point: 143-145°
MDL No.: MFCD04973752
Formel:
C₇H₆BrFO Molecular Weight: 205,03 g/mol Siedepunkt: 143…145 °C Dichte: 1,599 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973752 CAS-Nummer: 450-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...