2-Brom-5-fluoranisol

Lieferant: CHEMPUR
Achtung

Synonyme: 1-Bromo-4-fluoro-2-methoxybenzene

FL13776.POR 013776-25G
CHMPFL13776.POREA 0 EUR
CHMPFL13776.POR FLUO013776-25G
2-Brom-5-fluoranisol
2-Brom-5-fluoranisol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD04973752
CAS-Nummer: 450-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR