4-Brom-2,5-difluorphenol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H26212.06
H26212.14
H26212.06EA
78.7
EUR
H26212.06
H26212.14
4-Brom-2,5-difluorphenol ≥99%
4-Brom-2,5-difluorphenol
Purity: 99%
CAS No.: 486424-36-6
Molecular Formula: C6H3BrF2O
Formula Weight: 208.99
Melting Point: 51-53°
MDL No.: MFCD04973751
Formel:
C₆H₃BrF₂O Molecular Weight: 208.99 g/mol Schmelzpunkt: 51…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973751 CAS-Nummer: 486424-36-6 EINECS: None |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...