4-Brom-2,5-difluorphenol

Lieferant: CHEMPUR
Achtung

FL13775.POR 013775-25G 013775-5G
CHMPFL13775.POREA 0 EUR
CHMPFL13775.POR FLUO013775-25G FLUO013775-5G
4-Brom-2,5-difluorphenol
4-Brom-2,5-difluorphenol
Formel: C₆H₃BrF₂O
Molecular Weight: 208,99 g/mol
MDL: MFCD04973751
CAS-Nummer: 486424-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR