4-Brom-3-fluorphenol

Lieferant: FLUOROCHEM

019215-25G 019215-100G
FLUO019215-25GEA 130 EUR
FLUO019215-25G FLUO019215-100G
4-Brom-3-fluorphenol
4-Brom-3-fluorphenol
Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
MDL: MFCD00051907
CAS-Nummer: 121219-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR