2-Bromphenylisocyanat

Lieferant: Thermo Scientific

Synonyme: Isocyansäure-2-bromphenylester

TL00795.1 TL00795.10
MAYBTL00795.1EA 0 EUR
MAYBTL00795.1 MAYBTL00795.10
2-Bromphenylisocyanat
2-Bromphenylisocyanat
Formel: C₇H₄BrNO
Molecular Weight: 198.02 g/mol
Siedepunkt: 62…64 °C (0.5 mmHg)
Dichte: 1.6 g/cm³ (20 °C)
MDL: MFCD00001995
CAS-Nummer: 1592-00-3
UN: 2206
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR