2,6-Dibrom-4-nitrophenol

Lieferant: CHEMPUR

Synonyme: 1,3-Dibromo-2-hydroxy-5-nitrobenzene

RDR01637.250 RDR01637.1
CHMPRDR01637.250EA 0 EUR
CHMPRDR01637.250 MAYBRDR01637.1
2,6-Dibrom-4-nitrophenol
2,6-Dibrom-4-nitrophenol
Formel: C₆H₃Br₂NO₃
Schmelzpunkt: 148,5 °C
MDL: MFCD00007334
CAS-Nummer: 99-28-5
EINECS: 202-744-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR