4-Brombutylphenylether ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Phenoxybutylbromid, 1-Brom-4-phenoxybutan

A14956.06 A14956.14 A14956.22
A14956.06EA 25.7 EUR
A14956.06 A14956.14 A14956.22
4-Brombutylphenylether ≥97%
4-Brombutylphenylether

Purity: 97% CAS No.: 1200-03-9 Molecular Formula: C6H5O(CH2)4Br Formula Weight: 229.12 Flash Point: 110°(230°F) Odour: Odourless Boiling Point: 153-154°/18mm Beilstein Reference: 2044374 MDL No.: MFCD00000262

Formel: (C₆H₅)O(CH₂)₄Br
Molecular Weight: 229,12 g/mol
Siedepunkt: 153…154 °C (18 mmHg)
Schmelzpunkt: 40…43 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000262
CAS-Nummer: 1200-03-9
EINECS: 214-849-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR